Anti-DP1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB43490-100EA 0 EUR
BIRBORB43490-100
Anti-DP1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: DP1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR