Anti-GFI1B Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB43502-100EA 0 EUR
BIRBORB43502-100
Anti-GFI1B Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: GFI1B (growth factor independent 1B transcription repressor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR