Anti-Intra-Acrosomal Protein Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB43877-0.1EA 0 EUR
BIRBORB43877-0.1
Anti-Intra-Acrosomal Protein Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: Intra-Acrosomal Protein
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR