Anti-ITGA2B Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB44000-100EA 0 EUR
BIRBORB44000-100
Anti-ITGA2B Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: ITGA2B (integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigen CD41))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR