Anti-CD59 Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB44058-0.1EA 0 EUR
BIRBORB44058-0.1
Anti-CD59 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: CD59 (CD59 molecule, complement regulatory protein)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR