Imprimer

Anti-CD14 Mouse Monoclonal Antibody

Fournisseur: Biorbyt
BIRBORB44148-100EA 0 EUR
BIRBORB44148-100
Anti-CD14 Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: CD14 (CD14 molecule)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR