Anti-IGF2R Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB44200-100EA 0 EUR
BIRBORB44200-100
Anti-IGF2R Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: IGF2R (insulin-like growth factor 2 receptor)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR