Anti-CD4 Mouse Monoclonal Antibody (Peridinin Chlorophyll)

Supplier: Biorbyt
BIRBORB44212-100EA 0 EUR
BIRBORB44212-100
Anti-CD4 Mouse Monoclonal Antibody (Peridinin Chlorophyll)
Antibodies
Type: Primary
Antigen: CD4 (CD4 molecule)
Clonality: monoclonal
Clone:
Conjugation: Peridinin Chlorophyll
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR