Imprimer

Anti-HLA-E Mouse Monoclonal Antibody

Fournisseur: Biorbyt
BIRBORB44421-0.1EA 0 EUR
BIRBORB44421-0.1
Anti-HLA-E Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: HLA-E (major histocompatibility complex, class I, E)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR