Anti-Tenascin Mouse Monoclonal Antibody
Supplier: Biorbyt
BIRBORB44503-0.1EA 0 EUR
BIRBORB44503-0.1
Anti-Tenascin Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: Tenascin
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human