Anti-CD36 Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB44516-0.1EA 0 EUR
BIRBORB44516-0.1
Anti-CD36 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: CD36 (CD36 molecule (thrombospondin receptor))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR