Anti-TNFRSF10C Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB44520-0.1EA 0 EUR
BIRBORB44520-0.1
Anti-TNFRSF10C Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: TNFRSF10C (tumor necrosis factor receptor superfamily, member 10c, decoy without an intracellular domain)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR