Imprimer

Anti-TUBB Mouse Monoclonal Antibody

Fournisseur: Biorbyt
BIRBORB44535-0.1EA 0 EUR
BIRBORB44535-0.1
Anti-TUBB Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: TUBB (tubulin, beta class I)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR