Anti-MS4A1 Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB44641-0.1EA 0 EUR
BIRBORB44641-0.1
Anti-MS4A1 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: MS4A1 (membrane-spanning 4-domains, subfamily A, member 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR