Anti-STRO-1 Mouse Monoclonal Antibody
Supplier: Biorbyt
BIRBORB44752-0.1EA 0 EUR
BIRBORB44752-0.1
Anti-STRO-1 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: STRO-1
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human