Anti-CD33 Mouse Monoclonal Antibody (FITC (Fluorescein))
Supplier: Biorbyt
BIRBORB44760-100EA 0 EUR
BIRBORB44760-100
Anti-CD33 Mouse Monoclonal Antibody (FITC (Fluorescein))
Antibodies
Type: Primary
Antigen: CD33 (CD33 molecule)
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now