Anti-LIME1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB44779-0.1EA 0 EUR
BIRBORB44779-0.1
Anti-LIME1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: LIME1 (Lck interacting transmembrane adaptor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR