Anti-CD9 Rat Monoclonal Antibody (FITC (Fluorescein))

Supplier: Biorbyt
BIRBORB44805-0.1EA 0 EUR
BIRBORB44805-0.1
Anti-CD9 Rat Monoclonal Antibody (FITC (Fluorescein))
Antibodies
Type: Primary
Antigen: CD9 (CD9 molecule)
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR