Anti-Midkine Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB47997-100EA 0 EUR
BIRBORB47997-100
Anti-Midkine Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Midkine
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR