Anti-EDA Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB48027-100EA 0 EUR
BIRBORB48027-100
Anti-EDA Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: EDA (ectodysplasin A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR