Anti-ALDH1A1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB48279-100EA 0 EUR
BIRBORB48279-100
Anti-ALDH1A1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ALDH1A1 (aldehyde dehydrogenase 1 family, member A1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR