Anti-ANXA2 Rabbit Polyclonal Antibody
Supplier: Biorbyt
BIRBORB48281-100EA 0 EUR
BIRBORB48281-100
Anti-ANXA2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ANXA2 (annexin A2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat