Anti-USP28 Rabbit Polyclonal Antibody
: Biorbyt
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)
Type: Primary
Antigen: USP28 (ubiquitin specific peptidase 28)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
- Catalog No:
- BIRBORB100097-200
- BIRBORB100097-100
- BIRBORB100097-5
- BIRBORB100097-20
- Symbole de l'antigène:
- USP28
- USP28
- USP28
- USP28
- Nom de l'antigène:
- Ubiquitin specific peptidase 28
- Ubiquitin specific peptidase 28
- Ubiquitin specific peptidase 28
- Ubiquitin specific peptidase 28
- Type d'anticorps:
- Primaire
- Primaire
- Primaire
- Primaire
- Clonalité:
- Polyclonal
- Polyclonal
- Polyclonal
- Polyclonal
- Conjugaison:
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Réactivité:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Human, Rat, Mouse
- Human, Rat, Mouse
- Hôte:
- Rabbit
- Rabbit
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- IgG
- IgG
- Western blot:
- Yes
- Yes
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Yes
- Yes
- ImmunoChimie:
- Yes
- Yes
- Yes
- Yes
- Formulaire:
- liquid
- liquid
- liquid
- liquid
- Molecular weight:
- 100 kDa
- 100 kDa
- 100 kDa
- 100 kDa
- Purification:
- purified by affinity chromatography
- purified by affinity chromatography
- purified by affinity chromatography
- purified by affinity chromatography
- Concentration:
- 0,5 mg/ml
- 0,5 mg/ml
- 0,05 mg/ml
- 0,20 mg/ml
- Storage buffer:
- 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Température de stockage:
- Cool
- Cool
- Cool
- Cool
- Température de transport:
- Ice
- Ice
- Ice
- Ice
- TailleCdt
Specifications
Symbole de l'antigèneUSP28Nom de l'antigèneUbiquitin specific peptidase 28Environmentally PreferableType d'anticorpsPrimaireClonalitéPolyclonalConjugaisonUnconjugatedRéactivitéHuman, Rat, MouseHôteRabbitIsotypeIgGWestern blotYesELISAYesImmunoChimieYesFormulaireliquidMolecular weight100 kDaPurificationpurified by affinity chromatographyConcentration0,5 mg/mlStorage buffer10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerolTempérature de stockageCoolTempérature de transportIceSpecifications
Symbole de l'antigèneUSP28Nom de l'antigèneUbiquitin specific peptidase 28Environmentally PreferableType d'anticorpsPrimaireClonalitéPolyclonalConjugaisonUnconjugatedRéactivitéHuman, Rat, MouseHôteRabbitIsotypeIgGWestern blotYesELISAYesImmunoChimieYesFormulaireliquidMolecular weight100 kDaPurificationpurified by affinity chromatographyConcentration0,5 mg/mlStorage buffer10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerolTempérature de stockageCoolTempérature de transportIceSpecifications
Symbole de l'antigèneUSP28Nom de l'antigèneUbiquitin specific peptidase 28Environmentally PreferableType d'anticorpsPrimaireClonalitéPolyclonalConjugaisonUnconjugatedRéactivitéHuman, Rat, MouseHôteRabbitIsotypeIgGWestern blotYesELISAYesImmunoChimieYesFormulaireliquidMolecular weight100 kDaPurificationpurified by affinity chromatographyConcentration0,05 mg/mlStorage buffer10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerolTempérature de stockageCoolTempérature de transportIceSpecifications
Symbole de l'antigèneUSP28Nom de l'antigèneUbiquitin specific peptidase 28Environmentally PreferableType d'anticorpsPrimaireClonalitéPolyclonalConjugaisonUnconjugatedRéactivitéHuman, Rat, MouseHôteRabbitIsotypeIgGWestern blotYesELISAYesImmunoChimieYesFormulaireliquidMolecular weight100 kDaPurificationpurified by affinity chromatographyConcentration0,20 mg/mlStorage buffer10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerolTempérature de stockageCoolTempérature de transportIce