Anti-VCAM1 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB115135-100EA 510 EUR
ABCAAB115135-100
Anti-VCAM1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: VCAM1 (vascular cell adhesion molecule 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR