Anti-COMMD4 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB115169-100EA 510 EUR
ABCAAB115169-100
Anti-COMMD4 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: COMMD4 (COMM domain containing 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR