Imprimer

Anti-ZBTB1 Rabbit Polyclonal Antibody

Fournisseur: Abcam
ABCAAB115170-100EA 510 EUR
ABCAAB115170-100
Anti-ZBTB1 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: ZBTB1 (zinc finger and BTB domain containing 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR