Anti-BRCC3 Rabbit polyclonal antibody unconjugated

Supplier: Abcam
ABCAAB115172-100EA 550 EUR
ABCAAB115172-100
Anti-BRCC3 Rabbit polyclonal antibody unconjugated
Antibodies
Type: Primary
Antigen: BRCC3 (BRCA1/BRCA2-containing complex, subunit 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR