Anti-LEFTY2 Mouse Monoclonal Antibody [clone: 7C5G1H6H10]

Supplier: Abcam
ABCAAB115224-50EA 530 EUR
ABCAAB115224-50
Anti-LEFTY2 Mouse Monoclonal Antibody [clone: 7C5G1H6H10]
Antibodies
Type: Primary
Antigen: LEFTY2 (left-right determination factor 2)
Clonality: monoclonal
Clone: 7C5G1H6H10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR