Anti-AP3 Goat Polyclonal Antibody

Supplier: Abcam
ABCAAB115234-50EA 550 EUR
ABCAAB115234-50
Anti-AP3 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: AP3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR