Anti-CCKAR Rabbit polyclonal antibody unconjugated

Supplier: Abcam
ABCAAB115287-50EA 550 EUR
ABCAAB115287-50
Anti-CCKAR Rabbit polyclonal antibody unconjugated
Antibodies
Type: Primary
Antigen: CCKAR (cholecystokinin A receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR