Anti-NLRC4 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB115537-50EA 480 EUR
ABCAAB115537-50
Anti-NLRC4 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: NLRC4 (NLR family, CARD domain containing 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR