Anti-NMUR2 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB115549-50EA 510 EUR
ABCAAB115549-50
Anti-NMUR2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: NMUR2 (neuromedin U receptor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR