Anti-ZIC5 Goat Polyclonal Antibody

Supplier: Abcam
ABCAAB115566-50EA 0 EUR
ABCAAB115566-50
Anti-ZIC5 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ZIC5 (Zic family member 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR