Anti-SLC12A4 Goat Polyclonal Antibody

Supplier: Abcam
ABCAAB115607-50EA 510 EUR
ABCAAB115607-50
Anti-SLC12A4 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SLC12A4 (solute carrier family 12 (potassium/chloride transporter), member 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR