Anti-CD14 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB115634-100EA 550 EUR
ABCAAB115634-100
Anti-CD14 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CD14 (CD14 molecule)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR