Imprimer

Anti-DRD3 Rabbit Polyclonal Antibody

Fournisseur: Abcam
ABCAAB115675-50EA 550 EUR
ABCAAB115675-50
Anti-DRD3 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: DRD3 (dopamine receptor D3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR