Anti-HSF1 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB115702-50EA 550 EUR
ABCAAB115702-50
Anti-HSF1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: HSF1 (heat shock transcription factor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR