Anti-CASP9 Mouse monoclonal antibody unconjugated [clone: 10-1-87]

Supplier: Abcam
ABCAAB115792-100EA 525 EUR
ABCAAB115792-100
Anti-CASP9 Mouse monoclonal antibody unconjugated [clone: 10-1-87]
Antibodies
Type: Primary
Antigen: CASP9 (caspase 9, apoptosis-related cysteine peptidase)
Clonality: monoclonal
Clone: 32051
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR