Anti-FGF10 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB115825-100EA 550 EUR
ABCAAB115825-100
Anti-FGF10 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: FGF10 (fibroblast growth factor 10)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR