Anti-PTP rho Goat Polyclonal Antibody

Supplier: Abcam
ABCAAB115848-50EA 510 EUR
ABCAAB115848-50
Anti-PTP rho Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PTP rho
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR