Anti-Hsp30 Mouse monoclonal antibody [clone: RT30.1]

Supplier: Abcam
ABCAAB115864-50EA 455 EUR
ABCAAB115864-50
Anti-Hsp30 Mouse monoclonal antibody [clone: RT30.1]
Type: Primary
Antigen: Hsp30
Clonality: monoclonal
Clone: RT30.1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Fish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR