Anti-PKM Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB116271-100EA 460 EUR
ABCAAB116271-100
Anti-PKM Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PKM (pyruvate kinase, muscle)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR