Anti-GPR146 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB116585-50EA 550 EUR
ABCAAB116585-50
Anti-GPR146 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: GPR146 (G protein-coupled receptor 146)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR