Anti-KEAP1 Goat Polyclonal Antibody

Supplier: Abcam
ABCAAB118285-200EA 0 EUR
ABCAAB118285-200
Anti-KEAP1 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: KEAP1 (kelch-like ECH-associated protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR