Anti-KRT14 Rabbit Monoclonal Antibody [clone: SP53]

Supplier: Abcam
ABCAAB119695-100EA 540 EUR
ABCAAB119695-100
Anti-KRT14 Rabbit Monoclonal Antibody [clone: SP53]
Antibodies
Type: Primary
Antigen: KRT14 (keratin 14)
Clonality: monoclonal
Clone: SP53
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR