Anti-CDH5 Rabbit polyclonal antibody unconjugated

Supplier: Abcam
ABCAAB119785-100EA 575 EUR
ABCAAB119785-100
Anti-CDH5 Rabbit polyclonal antibody unconjugated
Antibodies
Type: Primary
Antigen: CDH5 (cadherin 5, type 2 (vascular endothelium))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR