Anti-PDGFRB Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 18A2]

Supplier: Abcam
ABCAAB119861-50EA 550 EUR
ABCAAB119861-50
Anti-PDGFRB Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 18A2]
Antibodies
Type: Primary
Antigen: PDGFRB (platelet-derived growth factor receptor, beta polypeptide)
Clonality: monoclonal
Clone: 18A2
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR