Anti-FGG Mouse Monoclonal Antibody [clone: 5A6]

Supplier: Abcam
ABCAAB119948-100EA 575 EUR
ABCAAB119948-100
Anti-FGG Mouse Monoclonal Antibody [clone: 5A6]
Antibodies
Type: Primary
Antigen: FGG (fibrinogen gamma chain)
Clonality: monoclonal
Clone: 5A6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR