Anti-STMN1 Rabbit Monoclonal Antibody [clone: EP1572]

Supplier: Abcam
ABCAAB119991-100EA 560 EUR
ABCAAB119991-100
Anti-STMN1 Rabbit Monoclonal Antibody [clone: EP1572]
Antibodies
Type: Primary
Antigen: STMN1 (stathmin 1)
Clonality: monoclonal
Clone: EP1572
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR