Anti-NMNAT3 Goat Polyclonal Antibody

Supplier: Abcam
ABCAAB121030-200EA 550 EUR
ABCAAB121030-200
Anti-NMNAT3 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: NMNAT3 (nicotinamide nucleotide adenylyltransferase 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR